(Methoxycarbonylmethyl)triphenylphosphoniumbromid

Lieferant: Molekula

MOLEM89985323EA 0 EUR
MOLEM89985323
(Methoxycarbonylmethyl)triphenylphosphoniumbromid
(Methoxycarbonylmethyl)triphenylphosphoniumbromid
Formel: C₂₁H₂₀BrO₂P
Molecular Weight: 415,28 g/mol
MDL: MFCD00011801
CAS-Nummer: 1779-58-4
EINECS: 217-222-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR