(R)-(+)-Pulegon, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ quality level = MQ100

SIALP55708-100MLEA 123 EUR
SIALP55708-100ML
(R)-(+)-Pulegon, Sigma-Aldrich®
(R)-(+)-Pulegon
Formel: C₁₀H₁₆O
Molecular Weight: 152,23 g/mol
MDL: MFCD00063000
CAS-Nummer: 89-82-7
EINECS: 201-943-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR