3-Methoxy-2-(trifluormethyl)benzaldehyd ≥97%

Lieferant: Thermo Scientific
Achtung

H33018.03EA 92.3 EUR
H33018.03 H33018.MD
3-Methoxy-2-(trifluormethyl)benzaldehyd ≥97%
3-Methoxy-2-(trifluoromethyl)benzaldehyde
Formel: C₉H₇F₃O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD13185677
CAS-Nummer: 1214384-89-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR