2-Piperazinon ≥97%
Lieferant: Thermo Scientific
|
Achtung
|
B24131.03EA
67.3
EUR
B24131.03
B24131.14
B24131.06
2-Piperazinon ≥97%
2-Piperazinon
Formel:
C₄H₈N₂O Molecular Weight: 100,12 Schmelzpunkt: 129…134 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD01318687 CAS-Nummer: 5625-67-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...