4'-Morpholinoacetophenon, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ quality level = MQ100

SIAL119865-5GEA 36.3 EUR
SIAL119865-5G
4'-Morpholinoacetophenon, Sigma-Aldrich®
4'-Morpholinoacetophenon
Formel: C₁₂H₁₅NO₂
MDL: MFCD00006168
CAS-Nummer: 39910-98-0
EINECS: 254-698-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR