3-Chlorisatosäureanhydrid, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ quality level = MQ100

SIAL691739-1GEA 88 EUR
SIAL691739-1G
3-Chlorisatosäureanhydrid, Sigma-Aldrich®
3-Chlorisatosäureanhydrid
Formel: C₈H₄ClNO₃
MDL: MFCD06809785
CAS-Nummer: 63497-60-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR