2,3,4-Trifluorrtoluol, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ quality level = MQ100

SIAL340472-1GEA 120 EUR
SIAL340472-1G
2,3,4-Trifluorrtoluol, Sigma-Aldrich®
2,3,4-Trifluorrtoluol
Formel: C₇H₅F₃
Molecular Weight: 146,1098 g/mol
MDL: MFCD00075194
CAS-Nummer: 193533-92-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR