4-Fluor-3-nitrophenylboronsäure

Lieferant: COMBI-BLOCKS INC.

COBBBB-2049-1GEA 159 EUR
COBBBB-2049-1G
4-Fluor-3-nitrophenylboronsäure
4-Fluor-3-nitrophenylboronsäure
Formel: C₆H₅BFNO₄
Molecular Weight: 184,9176 g/mol
MDL: MFCD08277236
CAS-Nummer: 352530-22-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR