4-Phenoxyphenylboronsäure, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ quality level = MQ100

SIAL480142-5GEA 68.1 EUR
SIAL480142-5G
4-Phenoxyphenylboronsäure, Sigma-Aldrich®
4-Phenoxyphenylboronsäure
Formel: C₁₂H₁₁BO₃
Molecular Weight: 214,03 g/mol
MDL: MFCD00093312
CAS-Nummer: 51067-38-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR