4-Fluor-3-nitrophenylboronsäurepinakolester

Lieferant: COMBI-BLOCKS INC.

COBBPN-2049-250MEA 85.1 EUR
COBBPN-2049-250M COBBPN-2049-1G
4-Fluor-3-nitrophenylboronsäurepinakolester
4-Fluor-3-nitrophenylboronsäurepinakolester
Formel: C₁₂H₁₅BFNO₄
Molecular Weight: 267,06 g/mol
MDL: MFCD12546515
CAS-Nummer: 1218791-09-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR