5-Brom-2,4-difluorbenzaldehyd

Lieferant: COMBI-BLOCKS INC.

COBBOT-2081-1GEA 46.6 EUR
COBBOT-2081-1G
5-Brom-2,4-difluorbenzaldehyd
5-Brom-2,4-difluorbenzaldehyd
Formel: C₇H₃BrF₂O
MDL: MFCD09842439
CAS-Nummer: 473416-91-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR