4-(Methylsulfanyl)-1-butanol, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

SIAL319694-1GEA 53.8 EUR
SIAL319694-1G
4-(Methylsulfanyl)-1-butanol, Sigma-Aldrich®
4-(Methylthio)butanol
Formel: C₅H₁₂OS
Molecular Weight: 120,21 g/mol
MDL: MFCD00002973
CAS-Nummer: 20582-85-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR