2,3,4,4'-Tetrahydroxybenzophenon ≥98,0% (durch titrimetrische Analyse)

Achtung

TCIAT1594-25GEA 71 EUR
TCIAT1594-25G
2,3,4,4'-Tetrahydroxybenzophenon ≥98,0% (durch titrimetrische Analyse)
2,3,4,4'-Tetrahydroxybenzophenon
Formel: C₁₃H₁₀O₅
Molecular Weight: 246,2155 g/mol
MDL: MFCD00083097
CAS-Nummer: 31127-54-5
https://de.vwr.com/stibo/web/std.lang.all/56/01/28995601.jpg

Bestellen

Spezifikation Testergebnisse

Appearance Light yellow to Amber to Dark green powder to crystal
Purity (Neutralization titration) min. 98.0 %
Melting point 218.0 to 222.0 deg-C
Solubility in Methanol very faint turbidity

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR