4-Methylhippursäure, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ quality level = MQ100

SIAL328022-1GEA 54.3 EUR
SIAL328022-1G
4-Methylhippursäure, Sigma-Aldrich®
4-Methylhippursäure
Formel: C₁₀H₁₁NO₃
Molecular Weight: 193,20
MDL: MFCD00020449
CAS-Nummer: 27115-50-0
EINECS: 248-231-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR