1,3-Di(p-tolyl)-2-thioharnstoff, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ quality level = MQ100

SIAL131091-5GEA 90.4 EUR
SIAL131091-5G
1,3-Di(p-tolyl)-2-thioharnstoff, Sigma-Aldrich®
1,3-Di(p-tolyl)-2-thioharnstoff
Formel: C₁₅H₁₆N₂S
MDL: MFCD00008541
CAS-Nummer: 621-01-2
EINECS: 210-665-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR