1-Cetylpyridiniumbromid Monohydrat, Sigma-Aldrich®
Lieferant: Merck
M-Clarity™ quality level = MQ100
SIAL285315-100GEA
108
EUR
SIAL285315-100G
1-Cetylpyridiniumbromid Monohydrat, Sigma-Aldrich®
1-Cetylpyridiniumbromid Monohydrat
Formel:
C₂₁H₃₈BrN·1H₂O Molecular Weight: 402,47 g/mol (384,44 anhy) |
MDL:
MFCD00150003 CAS-Nummer: 202869-92-9 EINECS: 205-428-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...