2',4'-Dimethoxypropiophenon, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ quality level = MQ100

SIAL592137-5GEA 164 EUR
SIAL592137-5G
2',4'-Dimethoxypropiophenon, Sigma-Aldrich®
2',4'-Dimethoxypropiophenon
Formel: C₁₁H₁₄O₃
MDL: MFCD00027012
CAS-Nummer: 831-00-5
EINECS: 212-601-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR