Ethylchlorfluoracetat, Sigma-Aldrich®
Lieferant: Merck
M-Clarity™ quality level = MQ100
SIAL539147-25GEA
62.9
EUR
SIAL539147-25G
Ethylchlorfluoracetat, Sigma-Aldrich®
Ethylchlorfluoracetat
Formel:
C₄H₆ClFO₂ Molecular Weight: 140,54 g/mol |
MDL:
MFCD00039333 CAS-Nummer: 401-56-9 EINECS: 206-930-5 UN: 2920 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...