D(+)-Turanose
Lieferant: EHRENSTORFER GMBH
EHERC17895500EA
45
EUR
EHERC17895500
D(+)-Turanose
D(+)-Turanose
Formel:
C₁₂H₂₂O₁₁ Molecular Weight: 342,30 g/mol |
MDL:
MFCD00006606 CAS-Nummer: 547-25-1 EINECS: 208-918-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...