5-Fluor-2-(trifluormethoxy)anilin ≥97%

Lieferant: Thermo Scientific
Gefahr

H33278.MDEA 33.9 EUR
H33278.MD H33278.03
5-Fluor-2-(trifluormethoxy)anilin ≥97%
5-Fluor-2-(trifluormethoxy)anilin
Formel: C₇H₅F₄NO
Molecular Weight: 195,11
Lagertemperatur: Raumtemperatur
MDL: MFCD16652514
CAS-Nummer: 123572-63-4
UN: 2810
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR