3,4-Difluor-o-toluylsäure ≥97%

Lieferant: Thermo Scientific
Achtung

H32326.03EA 50.8 EUR
H32326.03 H32326.06
3,4-Difluor-o-toluylsäure ≥97%
3,4-Difluor-o-toluylsäure
Formel: C₈H₆F₂O₂
Molecular Weight: 172,13
Schmelzpunkt: 152…156 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD07777153
CAS-Nummer: 157652-31-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR