Undecyltrichlorsilan

Lieferant: SILANE (PART OF CHEMPUR)

SILNSILS21650.10EA 0 EUR
SILNSILS21650.10
Undecyltrichlorsilan
Undecyltrichlorsilan
Formel: C₁₁H₂₃Cl₃Si
CAS-Nummer: 18052-07-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR