Trichlor(2,4,4-trimethylpentyl)silan

Lieferant: SILANE (PART OF CHEMPUR)

SILNSILS10225.25EA 167 EUR
SILNSILS10225.25
Trichlor(2,4,4-trimethylpentyl)silan
Trichloro(2,4,4-trimethylpentyl)silane
Formel: C₈H₁₇Cl₃Si
CAS-Nummer: 18379-25-4
EINECS: 242-262-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR