Triacontyltrichlorsilan

Lieferant: SILANE (PART OF CHEMPUR)

SILNSILS16725.10EA 0 EUR
SILNSILS16725.10 SILNSILS16725.25 SILNSILS16725.100
Triacontyltrichlorsilan
Triacontyltrichlorosilane
Formel: C₃₀H₆₁Cl₃Si
CAS-Nummer: 70851-48-8
EINECS: 274-933-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR