Isobutyltrichlorsilan

Lieferant: SILANE (PART OF CHEMPUR)

SILNSILS10125.100EA 223 EUR
SILNSILS10125.100 SILNSILS10125.25
Isobutyltrichlorsilan
Isobutyltrichlorsilan
Formel: C₄H₉Cl₃Si
MDL: MFCD00053185
CAS-Nummer: 18169-57-8
EINECS: 242-053-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR