2-Propoxyphenylboronsäure

Lieferant: COMBI-BLOCKS INC.

COBBBB-8086-1GEA 83.1 EUR
COBBBB-8086-1G
2-Propoxyphenylboronsäure
2-Propoxyphenylboronsäure
Formel: C₉H₁₃BO₃
MDL: MFCD03427191
CAS-Nummer: 134896-34-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR