Kaliumhexafluorozirkonat, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ quality level = MQ100

SIAL336688-500GEA 60.5 EUR
SIAL336688-500G
Kaliumhexafluorozirkonat, Sigma-Aldrich®
Potassium hexafluorozirconate
Formel: F₆K₂Zr
Molecular Weight: 283,41
MDL: MFCD00042547
CAS-Nummer: 16923-95-8
EINECS: 240-985-6
UN: 3288
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR