4'-Methyl-3,4-dihydroxybenzophenon

Lieferant: U.S. Pharmacopeia

USPH1670218EA 1270 EUR
USPH1670218
4'-Methyl-3,4-dihydroxybenzophenon
4'-Methyl-3,4-dihydroxybenzophenon
Formel: C₁₄H₁₂O₃
EINECS: 000-000-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR