5'-Ethoxy-2'-hydroxyacetophenon, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ quality level = MQ100

SIAL579181-5GEA 132 EUR
SIAL579181-5G
5'-Ethoxy-2'-hydroxyacetophenon, Sigma-Aldrich®
5'-Ethoxy-2'-hydroxyacetophenon
Formel: C₁₀H₁₂O₃
MDL: MFCD05664258
CAS-Nummer: 56414-14-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR