2',3',4',5',6'-Pentamethyl-2-phenylacetophenon 95%

Achtung

H34080.09EA 140 EUR
H34080.09
2',3',4',5',6'-Pentamethyl-2-phenylacetophenon 95%
2',3',4',5',6'-Pentamethyl-2-phenylacetophenon
Formel: C₁₉H₂₂O
Molecular Weight: 266,38
Schmelzpunkt: 133…135 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00052403
CAS-Nummer: 147834-57-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR