Dimethylthexylchlorsilan

Lieferant: SILANE (PART OF CHEMPUR)

SILNSILS16525.25EA 0 EUR
SILNSILS16525.25 SILNSILS16525.10
Dimethylthexylchlorsilan
Dimethylthexylchlorsilan
Formel: C₈H₁₉ClSi
MDL: MFCD00012175
CAS-Nummer: 67373-56-2
UN: 2986
ADR: 8,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR