Dimethylthexylchlorsilan

Lieferant: ABCR GMBH

ABCRAB111427.0025EA 0 EUR
ABCRAB111427.0025
Dimethylthexylchlorsilan
Dimethylthexylchlorsilan
Formel: C₈H₁₉ClSi
MDL: MFCD00012175
CAS-Nummer: 67373-56-2
UN: 2986
ADR: 8,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR