Dichlordioctylsilan

Lieferant: SILANE (PART OF CHEMPUR)

SILNSILS07665.5EA 0 EUR
SILNSILS07665.5 SILNSILS07665.25
Dichlordioctylsilan
Dioctyldichlorosilane
Formel: C₁₆H₃₄Cl₂Si
MDL: MFCD01074472
CAS-Nummer: 18416-07-4
UN: 2987
ADR: 8,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR