Dichlormethyloctylsilan
Lieferant: SILANE (PART OF CHEMPUR)
SILNSILS12650.25EA
50.9
EUR
SILNSILS12650.25
SILNSILS12650.100
Dichlormethyloctylsilan
Dichlormethyloctylsilan
Formel:
C₉H₂₀Cl₂Si |
MDL:
MFCD00042606 CAS-Nummer: 14799-93-0 EINECS: 238-863-2 UN: 2987 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...