Anilin-[15N] (98% ¹⁵N), Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ quality level = MQ200

SIAL488100-1GEA 323 EUR
SIAL488100-1G
Anilin-[15N] (98% ¹⁵N), Sigma-Aldrich®
Anilin-[15N]
Formel: C₆H₇N
MDL: MFCD00084195
CAS-Nummer: 7022-92-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR