4-Cyclopentyl-5-pyrimidinboronsäure

Lieferant: COMBI-BLOCKS INC.

COBBBB-5030-1GEA 850 EUR
COBBBB-5030-1G
4-Cyclopentyl-5-pyrimidinboronsäure
4-Cyclopentyl-5-pyrimidineboronic acid
Formel: C₉H₁₃BN₂O₂
MDL: MFCD10699617
CAS-Nummer: 1072945-80-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR