Maltoheptaose

Lieferant: Serva

SERA28380.02EA 123 EUR
SERA28380.02 SERA28380.01
Maltoheptaose
Maltoheptaose
Formel: C₄₂H₇₂O₃₆
Molecular Weight: 1152,9995 g/mol
MDL: MFCD00064605
CAS-Nummer: 34620-78-5
EINECS: 252-119-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR