Phenyl-2-pyridylketonoxim, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ quality level = MQ100

SIAL301884-5GEA 73.5 EUR
SIAL301884-5G
Phenyl-2-pyridylketonoxim, Sigma-Aldrich®
Phenyl-2-pyridylketonoxim
Formel: C₁₂H₁₀N₂O
Molecular Weight: 198,22 g/mol
MDL: MFCD00006310
CAS-Nummer: 1826-28-4
EINECS: 217-374-8
UN: 2811
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR