2'-(4-Acetophenoxy)-3'-chlor-5'-(trifluormethyl)pyridin

Lieferant: MENAI (PART OF CHEMPUR)

MENACB470.5EA 0 EUR
MENACB470.5
2'-(4-Acetophenoxy)-3'-chlor-5'-(trifluormethyl)pyridin
2'-(4-Acetophenoxy)-3'-chlor-5'-(trifluormethyl)pyridin
Formel: C₁₄H₉CIF₃NO₂
MDL: MFCD00170137
CAS-Nummer: 217186-15-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR