5-Fluornicotinamid
Lieferant: COMBI-BLOCKS INC.
COBBPY-7933-250MEA
232
EUR
COBBPY-7933-250M
COBBPY-7933-1G
5-Fluornicotinamid
5-Fluornicotinamid
Formel:
C₆H₅FN₂O |
MDL:
MFCD00160352 CAS-Nummer: 70-58-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...