2-Phenoxynicotinamid

Lieferant: Thermo Scientific

MAYBBTB03139.50EA 0 EUR
MAYBBTB03139.50
2-Phenoxynicotinamid
2-Phenoxynicotinamid
Formel: C₁₂H₁₀N₂O₂
MDL: MFCD00052252
CAS-Nummer: 111950-69-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR