2-Methoxyisonicotinamid

Lieferant: COMBI-BLOCKS INC.

COBBPY-7030-1GEA 185 EUR
COBBPY-7030-1G COBBPY-7030-5G
2-Methoxyisonicotinamid
2-Methoxyisonicotinamid
Formel: C₇H₈N₂O₂
MDL: MFCD00174337
CAS-Nummer: 105612-50-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR