2-Bromisonicotinamid
Lieferant: COMBI-BLOCKS INC.
COBBCA-4201-5GEA
341
EUR
COBBCA-4201-5G
COBBCA-4201-25G
COBBCA-4201-1G
2-Bromisonicotinamid
2-Bromisonicotinamid
Formel:
C₆H₅BrN₂O Molecular Weight: 201,02 g/mol |
MDL:
MFCD00234156 CAS-Nummer: 29840-73-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...