Lanthan(III)isopropoxid 98%
|
Gefahr
|
ACRO383640050EA
293
EUR
ACRO383640050
ACRO383640010
Lanthan(III)isopropoxid 98%
Lanthan(III)isopropoxid
Formel:
C₉H₂₁LaO₃ Molecular Weight: 316,18 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00015305 CAS-Nummer: 19446-52-7 UN: 2925 ADR: 4.1,II |
Spezifikation Testergebnisse
Appearance | White to light yellow Powder |
Infrared spectrum | Conforms |
Titration Complexometric | ≥97.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...