Bariumchlorat Monohydrat, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ quality level = MQ100

SIAL244554-500GEA 55.4 EUR
SIAL244554-500G
Bariumchlorat Monohydrat, Sigma-Aldrich®
Bariumchlorat Monohydrat
Formel: BaCl₂O₆·1H₂O
Molecular Weight: 322,26 g/mol (304,24 anhy)
MDL: MFCD00149147
CAS-Nummer: 10294-38-9
EINECS: 236-760-7
UN: 1445
ADR: 5.1,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR