Trypanblau

Lieferant: CLIN-TECH

CTEH80127EA 0 EUR
CTEH80127
Trypanblau
Trypanblau
Formel: C₃₄H₂₄N₆Na₄O₁₄S₄
Molecular Weight: 960,82 g/mol
MDL: MFCD00003969
CAS-Nummer: 72-57-1
EINECS: 200-786-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR