6-Morpholino-3-pyridinamin

Lieferant: SIA ENAMINE

ENAMEN300-25991.1GEA 120 EUR
ENAMEN300-25991.1G
6-Morpholino-3-pyridinamin
6-Morpholino-3-pyridinamin
Formel: C₉H₁₃N₃O
Molecular Weight: 179,22
MDL: MFCD00119562
CAS-Nummer: 52023-68-4
EINECS: 257-613-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR