Resodiacetophenon

Lieferant: CHEMPUR

FLUO012078.5GEA 72.2 EUR
FLUO012078.5G CHMPFL12078.POR
Resodiacetophenon
Resodiacetophenon
Formel: C₁₀H₁₀O₄
MDL: MFCD00017655
CAS-Nummer: 2161-85-5
EINECS: 218-482-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR