2-Butoxy-3-formyl-5-methylphenylboronsäure

Lieferant: COMBI-BLOCKS INC.

COBBBB-8233-1GEA 341 EUR
COBBBB-8233-1G
2-Butoxy-3-formyl-5-methylphenylboronsäure
2-Butoxy-3-formyl-5-methylphenylboronsäure
Formel: C₁₂H₁₇BO₄
MDL: MFCD08276747
CAS-Nummer: 480424-51-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR