5-Chlor-2-(methylamino)benzophenon, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ quality level = MQ100

SIAL191353-10GEA 26.7 EUR
SIAL191353-10G
5-Chlor-2-(methylamino)benzophenon, Sigma-Aldrich®
5-Chlor-2-(methylamino)benzophenon
Formel: C₁₄H₁₂ClNO
Molecular Weight: 245,71
MDL: MFCD00008284
CAS-Nummer: 1022-13-5
EINECS: 213-822-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR